Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012132197.3 | XP_011987587.1 | 5 - 6 | 643 | 105 | |
XM_012132205.3 | XP_011987595.1 | 5 - 6 | 722 | 105 | ||
XM_027964786.1 | XP_027820587.1 | 5 - 6 | 905 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |