Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101108840 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011661.4 | XP_004011710.2 | 1 - 2 | 731 | 62 |
Gene Symbol: FAM83A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972926.1 | XP_027828727.1 | 1 - 2 | 731 | 62 |