Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEU3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016324.4 | XP_004016373.1 | 2 - 3 | 522 | 56 | |
XM_015101081.2 | XP_014956567.1 | 1 - 2 | 1123 | 56 | ||
XM_027979536.1 | XP_027835337.1 | 2 - 3 | 2434 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |