Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020222.4 | XP_004020271.1 | 2 - 3 | 798 | 101 | |
XM_027960509.1 | XP_027816310.1 | 3 - 4 | 1062 | 101 | ||
XM_027960510.1 | XP_027816311.1 | 3 - 4 | 1062 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |