Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYREN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968830.1 | XP_027824631.1 | 1 - 2 | 1092 | 99 | |
XM_027968831.1 | XP_027824632.1 | 2 - 3 | 400 | 99 | ||
XM_027968833.1 | XP_027824634.1 | 2 - 3 | 338 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |