Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC114113036 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964633.1 | XP_027820434.1 | 1 | 485 | 94 |
Gene Symbol: CHCHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961836.1 | XP_027817637.1 | 3 - 4 | 605 | 94 |