Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1H21orf91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002817.4 | XP_004002866.1 | 4 - 5 | 798 | 75 | |
XM_012187061.3 | XP_012042451.1 | 4 - 5 | 844 | 75 |