Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027980298.1 | XP_027836099.1 | 3 - 4 | 866 | 103 | |
XM_027980303.1 | XP_027836104.1 | 3 - 4 | 836 | 103 | ||
XM_027980311.1 | XP_027836112.1 | 3 - 4 | 827 | 103 | ||
XM_027980320.1 | XP_027836121.1 | 3 - 4 | 1041 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |