Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014924.3 | XP_004014973.1 | 2 - 3 | 184 | 95 | |
XM_027977651.1 | XP_027833452.1 | 2 - 3 | 98 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |