Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012181006.3 | XP_012036396.3 | 4 - 5 | 3582 | 104 | |
XM_027971647.1 | XP_027827448.1 | 4 - 5 | 3585 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |