Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958026.1 | XP_027813827.1 | 2 - 3 | 6439 | 80 | |
XM_027958027.1 | XP_027813828.1 | 3 - 4 | 662 | 80 |