Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004772.4 | XP_004004821.1 | 3 - 4 | 540 | 75 | |
XM_012140664.3 | XP_011996054.1 | 2 - 3 | 496 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |