Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015095779.2 | XP_014951265.2 | 6 - 7 | 1614 | 77 | |
XM_027979144.1 | XP_027834945.1 | 7 - 8 | 1488 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |