Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100841.2 | XP_014956327.1 | 5 - 6 | 1188 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |