Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15H11orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979708.1 | XP_027835509.1 | 7 - 8 | 862 | 63 | |
XM_027979710.1 | XP_027835511.1 | 7 - 8 | 862 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |