Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPHB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010709.3 | XP_004010758.3 | 1 - 2 | 208 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |