Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014449.4 | XP_004014498.1 | 2 - 3 | 346 | 104 | |
XM_012188945.3 | XP_012044335.1 | 2 - 3 | 312 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |