Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014449.4 | XP_004014498.1 | 5 - 6 | 1214 | 92 | |
XM_012188945.3 | XP_012044335.1 | 5 - 6 | 1180 | 92 |