Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012098890.3 | XP_011954280.1 | 3 - 4 | 1144 | 70 | |
XM_012098892.3 | XP_011954282.1 | 3 - 4 | 1082 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |