Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002985.4 | XP_004003034.1 | 4 - 5 | 591 | 72 | |
XM_012095901.3 | XP_011951291.1 | 4 - 5 | 500 | 72 | ||
XM_027956526.1 | XP_027812327.1 | 3 - 4 | 466 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |