Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015099645.2 | XP_014955131.1 | 2 - 3 | 784 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |