Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF75D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012107284.3 | XP_011962674.2 | 5 - 6 | 1198 | 89 | |
XM_012107288.3 | XP_011962678.2 | 5 - 6 | 1108 | 89 | ||
XM_012107291.3 | XP_011962681.2 | 5 - 6 | 1108 | 89 | ||
XM_027963174.1 | XP_027818975.1 | 5 - 6 | 1108 | 89 | ||
XM_027963177.1 | XP_027818978.1 | 4 - 5 | 681 | 89 | ||
XM_027963178.1 | XP_027818979.1 | 4 - 5 | 750 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |