Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSMEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008062.3 | XP_004008111.2 | 2 - 3 | 233 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |