Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006255.4 | XP_004006304.1 | 5 - 6 | 473 | 83 | |
XM_012174098.3 | XP_012029488.1 | 5 - 6 | 475 | 83 | ||
XM_012174099.3 | XP_012029489.1 | 5 - 6 | 506 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |