Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966690.1 | XP_027822491.1 | 4 - 5 | 2019 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |