Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977754.1 | XP_027833555.1 | 6 - 7 | 988 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |