Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104623.2 | XP_014960109.2 | 5 - 6 | 1145 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |