Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006917.4 | XP_004006966.1 | 8 - 9 | 1306 | 98 | |
XM_012175494.3 | XP_012030884.1 | 8 - 9 | 1335 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |