Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975976.1 | XP_027831777.1 | 4 - 5 | 1815 | 84 | |
XM_027975977.1 | XP_027831778.1 | 4 - 5 | 690 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |