Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015093423.2 | XP_014948909.1 | 6 - 7 | 538 | 61 | |
XM_015093424.2 | XP_014948910.1 | 5 - 6 | 461 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |