Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006876.4 | XP_004006925.3 | 1 - 2 | 190 | 107 | |
GenBank mRNA | KU641038.1 | - | 1 - 2 | 88 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |