Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006876.4 | XP_004006925.3 | 3 - 4 | 413 | 80 | |
GenBank mRNA | KU641038.1 | - | 3 - 4 | 311 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |