Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960115.1 | XP_027815916.1 | 8 - 9 | 1891 | 68 | |
XM_027960116.1 | XP_027815917.1 | 7 - 8 | 967 | 68 | ||
XM_027960117.1 | XP_027815918.1 | 8 - 9 | 1894 | 68 | ||
XM_027960119.1 | XP_027815920.1 | 8 - 9 | 1895 | 68 | ||
XM_027960120.1 | XP_027815921.1 | 7 - 8 | 1797 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |