Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002768.3 | XP_004002817.2 | 6 - 7 | 1504 | 100 | |
XM_012186326.2 | XP_012041716.2 | 6 - 7 | 1540 | 100 | ||
XM_027978278.1 | XP_027834079.1 | 6 - 7 | 719 | 100 | ||
XM_027978283.1 | XP_027834084.1 | 4 - 5 | 470 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |