Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC85A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005934.4 | XP_004005983.3 | 1 - 2 | 345 | 58 | |
XM_027966810.1 | XP_027822611.1 | 1 - 2 | 344 | 58 | ||
XM_027966811.1 | XP_027822612.1 | 1 - 2 | 342 | 58 | ||
XM_027966812.1 | XP_027822613.1 | 1 - 2 | 345 | 58 | ||
XM_027966814.1 | XP_027822615.1 | 1 - 2 | 346 | 58 | ||
XM_027966815.1 | XP_027822616.1 | 1 - 2 | 345 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |