Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006199.4 | XP_004006248.3 | 4 - 5 | 612 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |