Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972999.1 | XP_027828800.1 | 2 - 3 | 200 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |