Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979417.1 | XP_027835218.1 | 4 - 5 | 1165 | 80 | |
XM_027979418.1 | XP_027835219.1 | 4 - 5 | 1165 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |