Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019206.4 | XP_004019255.2 | 3 - 4 | 494 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |