Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004023347.4 | XP_004023396.4 | 2 - 3 | 544 | 61 | |
XM_027976212.1 | XP_027832013.1 | 2 - 3 | 544 | 61 |