Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012672.4 | XP_004012721.1 | 4 - 5 | 681 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |