Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006348.4 | XP_004006397.2 | 2 - 3 | 440 | 74 | |
XM_027967315.1 | XP_027823116.1 | 2 - 3 | 1051 | 74 | ||
XM_027967316.1 | XP_027823117.1 | 3 - 4 | 613 | 74 |