Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975145.1 | XP_027830946.1 | 7 - 8 | 1296 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |