Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013671.4 | XP_004013720.1 | 1 - 2 | 373 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |