Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022355.4 | XP_004022404.1 | 2 - 3 | 507 | 111 | |
XM_004022356.4 | XP_004022405.1 | 2 - 3 | 364 | 111 | ||
XM_012107356.3 | XP_011962746.1 | 2 - 3 | 406 | 111 | ||
XM_027962993.1 | XP_027818794.1 | 2 - 3 | 547 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |