Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003247.4 | XP_004003296.2 | 6 - 7 | 6536 | 93 | |
XM_012101344.3 | XP_011956734.2 | 7 - 8 | 6330 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |