Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012173604.3 | XP_012028994.2 | 2 - 3 | 198 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |