Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016178.4 | XP_004016227.2 | 2 - 3 | 352 | 97 | |
XM_012095822.3 | XP_011951212.2 | 2 - 3 | 352 | 97 | ||
XM_012095823.3 | XP_011951213.2 | 2 - 3 | 352 | 97 |