Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015095217.2 | XP_014950703.2 | 4 - 5 | 2034 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |