Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022232.4 | XP_004022281.1 | 7 - 8 | 962 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |